Cyclopentadienyltitanium trichloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C1994-1G C1994-5G
TCC1994-1GEA 69 USD
TCC1994-1G TCC1994-5G
Cyclopentadienyltitanium trichloride ≥98.0% (by titrimetric analysis)
Cyclopentadienyltitanium trichloride

CAS Number: 1270-98-0 Molecular Formula: C5H5Cl3Ti Molecular Weight: 219.31 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 210


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Materials Chemistry, Polymerization Catalysts Other Product Names / Synonyms: Trichloro(cyclopentadienyl)titanium(IV)

Formula: C₅H₅Cl₃Ti
Melting Pt: 210 °C
MDL Number: MFCD00003724
CAS Number: 1270-98-0
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28af89522e0195d3fc2d4712e6eb2429c34dfa6c&vid=28af89522e0195d3fc2d4712e6eb2429c34dfa6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR