4-(Chloromethyl)benzonitrile ≥97.0%

Supplier: TCI America

C1985-25G C1985-5G
TCC1985-25GEA 95 USD
TCC1985-25G TCC1985-5G
4-(Chloromethyl)benzonitrile ≥97.0%
4-(Chloromethyl)benzonitrile

CAS Number: 874-86-2 Molecular Formula: C8H6ClN Molecular Weight: 151.59 Purity/Analysis Method: 97.0% (GC,T) Form: Crystal Boiling point (°C): 263 Melting point (°C): 80


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals Other Product Names / Synonyms: 4-(Chloromethyl)benzonitrile, alpha-Chloro-p-tolunitrile

Formula: C₈H₆ClN
MW: 151.60 g/mol
Boiling Pt: 263 °C
Melting Pt: 80 °C
MDL Number: MFCD00019761
CAS Number: 874-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b85abf76919b41928d59a927fa3289ff7fac3f5&vid=2b85abf76919b41928d59a927fa3289ff7fac3f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR