Copper(I) cyanide ≥98.0%

Supplier: TCI America

Synonyms: Copper cyanide, Cuprous cyanide

C1952-25G C1952-300G
TCC1952-25GEA 33.63 USD
TCC1952-25G TCC1952-300G
Copper(I) cyanide ≥98.0%
Copper (I) cyanide

CAS Number: 544-92-3 Molecular Formula: CCuN Molecular Weight: 89.56 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 473

Formula: CCuN
Melting Pt: 473 °C
MDL Number: MFCD00010975
CAS Number: 544-92-3
UN: 1587
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43ce64b7b666092aa42260ad1ce8618b21bb6500&vid=43ce64b7b666092aa42260ad1ce8618b21bb6500&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR