(Bromodifluoromethyl)trimethylsilane ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 115262-01-6
MDL Number: MFCD18641931
Molecular Formula: C4H9BrF2Si
Molecular Weight: 203.10
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Difluoromethylating Agents
Other Product Names / Synonyms: Bromodifluoro(trimethylsilyl)methane
Formula:
C₄H₉BrF₂Si Boiling Pt: 108 °C Density: 1.31 (20 °C) |
MDL Number:
MFCD18641931 CAS Number: 115262-01-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...