Biochanin A ≥98.0%
Supplier: TCI America
Synonyms:
5,7-Dihydroxy-3-(4-methoxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-methoxyphenyl)chromen-4-one, 4'-Methoxy-5,7-dihydroxyisoflavone, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-4'-methoxyisoflavone
CAS Number: 491-80-5
MDL Number: MFCD00006839
Molecular Formula: C16H12O5
Molecular Weight: 284.27
Purity/Analysis Method:
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 5,7-Dihydroxy-4'-methoxyisoflavone, Genistein 4'-Methyl Ether
Formula:
C₁₆H₁₂O₅ Melting Pt: 217 °C |
MDL Number:
MFCD00006839 CAS Number: 491-80-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...