2-Bromobenzyl mercaptan ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromobenzylmercaptan, o-Bromotoluene-α-thiol

B3747-25G B3747-5G
TCB3747-25GEA 335 USD
TCB3747-25G TCB3747-5G
2-Bromobenzyl mercaptan ≥98.0%
2-Bromobenzyl mercaptan

CAS Number: 143888-85-1 MDL Number: MFCD04038925 Molecular Formula: C7H7BrS Molecular Weight: 203.10 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 82 Flash Point (°C): 110 Specific Gravity (20/20): 1.53


Product Categories: Synthetic Organic Chemistry, Thiols Other Product Names / Synonyms: 2-Bromobenzenemethanethiol, (2-Bromophenyl)methanethiol

Formula: C₇H₇BrS
MW: 203.1
Boiling Pt: 82 °C
Density: 1.53 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD04038925
CAS Number: 143888-85-1
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36fa4a6c35fe3f4ecef4b0a217a7838066f68a74&vid=36fa4a6c35fe3f4ecef4b0a217a7838066f68a74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR