2-Bromo-4-fluorothiophene 95,0
Supplier: TCI America
B3680-1G
B3680-200MG
TCB3680-1GEA
1248
USD
TCB3680-1G
TCB3680-200MG
2-Bromo-4-fluorothiophene 95,0
2-Bromo-4-fluorothiophene
CAS Number: 32431-65-5
Molecular Formula: C4H2BrFS
Molecular Weight: 181.02
Purity/Analysis Method:
Formula:
C₄H₂BrFS MW: 181.02 g/mol Boiling Pt: 60 °C Density: 1.80 (20 °C) |
CAS Number:
32431-65-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...