(+)-Benzotetramisole ≥97.0% (by GC)
Supplier: TCI America
CAS Number: 885051-07-0
Molecular Formula: C15H12N2S
Molecular Weight: 252.34
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Optical Resolving Agents, Optical Resolving Agents
Other Product Names / Synonyms: (2R)-2,3-Dihydro-2-phenylimidazo[2,1-b]benzothiazole, (+)-BTM
Formula:
C₁₅H₁₂N₂S Melting Pt: 93 °C |
MDL Number:
MFCD11112169 CAS Number: 885051-07-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...