3-Bromothiophenol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: m-Bromothiophenol, 3-Bromobenzenethiol, 1-Bromo-3-mercaptobenzene, 3-Bromobenzene-1-thiol

B2348-25G B2348-5G
TCB2348-25GEA 208.52 USD
TCB2348-25G TCB2348-5G
3-Bromothiophenol ≥98.0% (by GC)
3-Bromothiophenol

CAS Number: 6320-01-0 MDL Number: MFCD00009603 Molecular Formula: C6H5BrS Molecular Weight: 189.07 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 121 Flash Point (°C): 105 Specific Gravity (20/20): 1.60 Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-Bromothiophenol

Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 121 °C
Density: 1.60 (20 °C)
Flash Pt: 105 °C
MDL Number: MFCD00009603
CAS Number: 6320-01-0
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4565d74c6fde99940eaf77bd7a748cd3dc16b5bf&vid=93a405f76c563eb3eb387b699d44b455b6769bcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR