Bromomethyl methyl ether ≥95.0%

Supplier: TCI America

B2131-25G B2131-5G
TCB2131-25GEA 143.43 USD
TCB2131-25G TCB2131-5G
Bromomethyl methyl ether ≥95.0%
Bromomethyl methyl ether

CAS Number: 13057-17-5 MDL Number: MFCD00000171 Molecular Formula: C2H5BrO Molecular Weight: 124.97 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point (°C): 88 Flash Point (°C): 26 Specific Gravity (20/20): 1.58 Storage Temperature: 0-10°C


Other Product Names / Synonyms: Methoxymethyl Bromide

Formula: C₂H₅BrO
Boiling Pt: 88 °C
Density: 1.58 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00000171
CAS Number: 13057-17-5
UN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afc4684748f96fdbd507c5c05107ae5e86b167d3&vid=afc4684748f96fdbd507c5c05107ae5e86b167d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR