(±)-3-Bromocyclohexene ≥90.0% (by GC)
Supplier: TCI America
CAS Number: 1521-51-3
MDL Number: MFCD00013775
Molecular Formula: C6H9Br
Molecular Weight: 161.04
Purity/Analysis Method:
Other Product Names / Synonyms: 1,2,3,4-Tetrahydrobromobenzene
Formula:
C₆H₉Br Boiling Pt: 82 °C Density: 1.40 (20 °C) Flash Pt: 54 °C |
MDL Number:
MFCD00013775 CAS Number: 1521-51-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...