Anisomycin ≥97%

Supplier: TCI America

Synonyms: (2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate

A2613-100MG A2613-25MG
TCA2613-100MGEA 444.53 USD
TCA2613-100MG TCA2613-25MG
Anisomycin ≥97%
Anisomycin

Anisomycin also termed as Flagecidin, inhibitor and anoikis sensitizer that functions by decreasing FLIP protein synthesis.


  • White crystals


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antibiotic Ingredients Other Product Names / Synonyms: Flagecidin, (2R,3S,4S)-2-(4-Methoxybenzyl)-3,4-pyrrolidinediol 3-Acetate

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Freezer
MDL Number: MFCD00077650
CAS Number: 22862-76-6
UN: 3462
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=930a67963c1004b8c8365c224db6af7bf1eaa5de&vid=930a67963c1004b8c8365c224db6af7bf1eaa5de&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR