3-Amino-3-(4-chlorophenyl)propionic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
3-Amino-3-(4-chlorophenyl)propanoic acid, 3-Amino-3-(4-chlorophenyl)propionic Acid, 3-Amino-3-(p-chlorophenyl)propionic acid
A2429-1G
A2429-5G
TCA2429-1GEA
258
USD
TCA2429-1G
TCA2429-5G
3-Amino-3-(4-chlorophenyl)propionic acid ≥97.0% (by HPLC, titration analysis)
3-Amino-3-(4-chlorophenyl)propionic acid
CAS Number: 19947-39-8
MDL Number: MFCD00126370
Molecular Formula: C9H10ClNO2
Molecular Weight: 199.63
Purity/Analysis Method:
Formula:
C₉H₁₀ClNO₂ MW: 199.64 g/mol Melting Pt: 220 °C |
MDL Number:
MFCD00126370 CAS Number: 19947-39-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...