Adefovir dipivoxil ≥98.0%

Supplier: TCI America

Synonyms: Adefovir Dipivoxil, 9-(2[bis(Pivaloyloxymethoxy)phosphorylmethoxy]ethyl)adenine, 9-[2-[Bis(pivaloyloxymethoxy)phosphorylmethoxy]ethyl]adenine, ((((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphoryl)bis(oxy))bis(methylene) bis(2,2-dimethylpropanoate)

A2414-100MG A2414-1G
TCA2414-100MGEA 34 USD
TCA2414-100MG TCA2414-1G
Adefovir dipivoxil ≥98.0%
Adefovir dipivoxil

CAS Number: 142340-99-6 MDL Number: MFCD00869897 Molecular Formula: C20H32N5O8P Molecular Weight: 501.48 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 101 Storage Temperature: 0°C


Product Categories: Bioscience, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs Other Product Names / Synonyms: 9-[2-[Bis(pivaloyloxymethoxy)phosphorylmethoxy]ethyl]adenine

Formula: C₂₀H₃₂N₅O₈P
MW: 501.48 g/mol
Melting Pt: 101 °C
MDL Number: MFCD00869897
CAS Number: 142340-99-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f865d16c5f096effdaf64edff44d072f542c882e&vid=f865d16c5f096effdaf64edff44d072f542c882e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR