2-Allyloxytetrahydropyran ≥95.0% (by GC)
Supplier: TCI America
CAS Number: 4203-49-0
MDL Number: MFCD00154089
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method:
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Tetrahydro-2-allyloxy-2H-pyran
Formula:
C₈H₁₄O₂ Boiling Pt: 75 °C Density: 0.96 (20 °C) Flash Pt: 35 °C |
MDL Number:
MFCD00154089 CAS Number: 4203-49-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...