Adenosine-3',5'-cyclic monophosphate (cAMP) ≥99.0% (by HPLC)

Supplier: TCI America

Synonyms: 3',5'-cAMP, 3′,5′-Cyclic AMP, Adenosine 3′,5′-cyclophosphate, (4aR,6R,7R,7aS)-6-(6-Amino-9H-purin-9-yl)-2,7-dihydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinine 2-oxide

A2381-1G A2381-5G
TCA2381-1GEA 96.23 USD
TCA2381-1G TCA2381-5G
Adenosine-3',5'-cyclic monophosphate (cAMP) ≥99.0% (by HPLC)
Adenosine-3',5'-cyclic monophosphate (cAMP)

CAS Number: 60-92-4
MDL Number: MFCD00005845
Molecular Formula: C10H12N5O6P
Molecular Weight: 329.21
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 220
Specific rotation [a]20/D: -50 deg (C=0.7, H2O)
Storage Temperature: <0°C


Product Categories: Bioscience, Nucleotides and Their Analogs
Other Product Names / Synonyms: 3',5'-Cyclic AMP, 3',5'-cAMP

Formula: C₁₀H₁₂N₅O₆P
MW: 329.21 g/mol
Melting Pt: 220 °C
MDL Number: MFCD00005845
CAS Number: 60-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f42ec34c746613a0a74f4694401a1d0c130f8607&vid=f42ec34c746613a0a74f4694401a1d0c130f8607&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR