Amlodipine besylate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
CAS Number: 111470-99-6
MDL Number: MFCD00887594
Molecular Formula: C20H25ClN2O5
Molecular Weight: 567.05
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 200
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antihypertensive Ingredients, Voltage-dependent Calcium Channel (VDCC) Antagonists
Formula:
C₂₀H₂₅ClN₂O₅ Melting Pt: 200 °C |
MDL Number:
MFCD00887594 CAS Number: 111470-99-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...