Allylboronic acid pinacol ester ≥96.0% stabilised

Supplier: TCI America

Synonyms: 2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane

A2157-1G A2157-5G
TCA2157-1GEA 42 USD
TCA2157-1G TCA2157-5G
Allylboronic acid pinacol ester ≥96.0% stabilised
Allylboronic acid pinacol ester

(stabilized with Phenothiazine) CAS Number: 72824-04-5 MDL Number: MFCD00013347 Molecular Formula: C9H17BO2 Molecular Weight: 168.04 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Boiling point (°C): 35 Flash Point (°C): 46 Specific Gravity (20/20): 0.89 Storage Temperature: 0°C


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives Other Product Names / Synonyms: Allylboronic Acid Pinacol Ester

Formula: C₉H₁₇BO₂
MW: 168.05 g/mol
Boiling Pt: 35 °C
Density: 0.89 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00013347
CAS Number: 72824-04-5
UN: 3272
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f80e85317681f5448b796ca32f5e3acf23a2e83a&vid=f80e85317681f5448b796ca32f5e3acf23a2e83a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR