4'-Aminohexanophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2058-25G A2058-5G
TCA2058-25GEA 637 USD
TCA2058-25G TCA2058-5G
4'-Aminohexanophenone ≥98.0% (by GC, titration analysis)
4'-Aminohexanophenone

CAS Number: 38237-76-2 Molecular Formula: C12H17NO Molecular Weight: 191.27 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 85


Other Product Names / Synonyms: 4-Hexanoylaniline

Formula: C₁₂H₁₇NO
Melting Pt: 85 °C
CAS Number: 38237-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=017082d66cc9fbf464f5ba0391176a5fb43fbca8&vid=017082d66cc9fbf464f5ba0391176a5fb43fbca8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR