2-Chloro-p-toluidine ≥98.0%
Supplier: TCI America
Synonyms:
2-Chloro-4-methylaniline, 4-Amino-3-chlorotoluene
A1437-25G
A1437-5G
TCA1437-25GEA
115
USD
TCA1437-25G
TCA1437-5G
2-Chloro-p-toluidine ≥98.0%
2-Chloro-p-toluidine
CAS Number: 615-65-6
MDL Number: MFCD00007666
Molecular Formula: C7H8ClN
Molecular Weight: 141.60
Purity/Analysis Method:
Other Product Names / Synonyms: 4-Amino-3-chlorotoluene, 2-Chloro-p-toluidine
Formula:
C₇H₈ClN MW: 141.6 Boiling Pt: 224 Melting Pt: 7 Density: 1.16 (20 °C) Flash Pt: 99 °C |
MDL Number:
MFCD00007666 CAS Number: 615-65-6 UN: 3429 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...