Anti-RORg Armenian Hamster Monoclonal Antibody [clone: RORg2]

Supplier: Biolegend
646501 646502
646501-BLEA 165.5 USD
646501-BL 646502-BL
Anti-RORg Armenian Hamster Monoclonal Antibody [clone: RORg2]
Antibodies
Purified anti-RORg [RORg2]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 1-2 μg per lane. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: RORg
Clonality: Monoclonal
Clone: RORg2
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR