Anti-IgG Goat Polyclonal Antibody (CF™ 640R)

Supplier: Biotium

20086 20086-1
71003-942EA 148.85 USD
71003-942 10018-408
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™640R dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red fluorescent CF™640R (Ex/Em 642/662 nm) is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647, and has exceptional photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 640R
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR