Anti-IgG Goat Polyclonal Antibody (CF™ 640R)

Supplier: Biotium

20175 20175-1
97065-548EA 260.49 USD
97065-548 97065-550
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 640R
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR