Anti-IgY Donkey Polyclonal Antibody (CF™ 350)

Supplier: Biotium

20275 20275-1
71004-010EA 260.49 USD
71004-010 71004-012
Anti-IgY Donkey Polyclonal Antibody (CF™ 350)
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: CF™ 350
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR