Anti-CD4 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: GK1.5]

Supplier: Biolegend
100427 100428
100427-BLEA 128.74 USD
100427-BL 100428-BL
Anti-CD4 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: GK1.5]
Antibodies
Pacific Blue™ anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR