Anti-IL-10 Rat Monoclonal Antibody (Biotin) [clone: JES5-16E3]

Supplier: Biolegend
505003 505004
505003-BLEA 113.7 USD
505003-BL 505004-BL
Anti-IL-10 Rat Monoclonal Antibody (Biotin) [clone: JES5-16E3]
Antibodies
Biotin anti-mouse IL-10 [JES5-16E3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: ELISA Detection, ELISPOT Detection, ICFC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA or ELISPOT detection, the antibody should be titrated between 0.25-1.0 μg/ml to determine optimal conditions. To obtain a linear standard curve, serial dilutions of IL-10 recombinant protein ranging from 2000 to 15 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: JES5-16E3
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR