Anti-IL-7 Rat Monoclonal Antibody (Biotin) [clone: BVD10-11C10]

Supplier: Biolegend
506601 506602
506601-BLEA 132.9 USD
506601-BL 506602-BL
Anti-IL-7 Rat Monoclonal Antibody (Biotin) [clone: BVD10-11C10]
Antibodies
Biotin anti-human IL-7 [BVD10-11C10]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ELISA Detection, ELISPOT Detection; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection applications, a concentration range of 1-4 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-7 recombinant protein ranging from 500 to 4 pg/ml are recommended for each ELISA plate. For use as an ELISPOT detection antibody, a concentration range of 2-8 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-7
Clonality: Monoclonal
Clone: BVD10-11C10
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR