Anti-IL-1b Mouse Monoclonal Antibody (Biotin) [clone: H1b-98]

Supplier: Biolegend
511703 511704
511703-BLEA 132.9 USD
511703-BL 511704-BL
Anti-IL-1b Mouse Monoclonal Antibody (Biotin) [clone: H1b-98]
Antibodies
Biotin anti-human IL-1β [H1b-98]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Detection, ELISPOT Detection, IF; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay.  Clone H1b-98 is useful as the detection antibody in a sandwich ELISA when used in conjunction with the purified H1b-27 antibody (Cat. No. 511601 or 511604). For ELISA detection applications, a concentration range of 1-4 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-1β recombinant protein ranging from 2000 to 31.3 pg/ml are recommended for each ELISA plate. For use as an ELISPOT detection antibody, a concentration range of 2-8 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-1b
Clonality: Monoclonal
Clone: H1b-98
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR