Anti-IL-12 p40 Mouse Monoclonal Antibody (Biotin) [clone: C8.6]

Supplier: Biolegend
508801 508802
508801-BLEA 113.7 USD
508801-BL 508802-BL
Anti-IL-12 p40 Mouse Monoclonal Antibody (Biotin) [clone: C8.6]
Antibodies
Biotin anti-human IL-12/IL-23 p40 [C8.6]; Isotype: Mouse IgG1, κ; Reactivity: Human, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ELISA Detection, ELISPOT Detection; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection applications, a concentration range of 0.5-2.0 μg/ml is recommended.  To obtain a linear standard curve, serial dilutions of IL-12 p40 recombinant protein ranging from 4000 to 30 pg/ml are recommended for each ELISA plate.  To obtain a linear standard curve, serial dilutions of IL-12 p70 recombinant protein ranging from 1000 to 8 pg/ml are recommended for each ELISA plate.  For use as an ELISPOT detection antibody, a concentration range of 1-4 μg/ml is recommended.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-12 p40
Clonality: Monoclonal
Clone: C8.6
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;IL-12/IL-23 p40 subunit (monomer;homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR