Anti-CD45RO Mouse Monoclonal Antibody [clone: UCHL-1]

Supplier: Diagnostic Biosystems

MOB043-05 MOB043
95024-826EA 284.79 USD
95024-826 99990-202
Anti-CD45RO Mouse Monoclonal Antibody [clone: UCHL-1]
Antibodies
Type:
Antigen: CD45RO
Clonality: Monoclonal
Clone: UCHL-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR