Rhenium ≥99.95% (trace metals basis), foil, Thickness 0.1 mm

Supplier: BeanTown Chemical

225855-25X25MM 225855-50X50MM
BT225855-25X25EA 209 USD
BT225855-25X25 BT225855-50X50
Rhenium ≥99.95% (trace metals basis), foil, Thickness 0.1 mm
Rhenium

CAS: 7440-15-5; EC No: 231-124-5; MDL No: MFCD00011195 Solid; Molecular Formula: Re; MW: 186.21 Melting Point: 3180°; Boiling Point: 5596° Density (g/mL): 21.02


  • Solid

Formula: Re
Boiling Pt: 5596 °C
Melting Pt: 3180 °C
Density: 21.02 g/mL
MDL Number: MFCD00011195
CAS Number: 7440-15-5
EINECS: 231-124-5

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR