Nickel ≥99.95% (trace metals basis), slug, Length 6.35 mm, Diameter 6.35 mm
Supplier: BeanTown Chemical
225595-1SLUG
225595-25SLUG
225595-5SLUG
BT225595-1SLUGEA
21
USD
BT225595-1SLUG
BT225595-25SLU
BT225595-5SLUG
Nickel ≥99.95% (trace metals basis), slug, Length 6.35 mm, Diameter 6.35 mm
Nickel
CAS: 7440-02-0; EC No: 231-111-4; MDL No: MFCD00011137
Solid; Molecular Formula: Ni; MW: 58.69
Melting Point: 1453°; Boiling Point: 2732°
Density (g/mL): 8.9
- Solid
Formula:
Ni Boiling Pt: 2732 °C Melting Pt: 1453 °C Density: 8.9 g/mL |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...