Strontium carbonate ≥99%

Supplier: BeanTown Chemical

224785-1KG 224785-250G
BT224785-1KGEA 124 USD
BT224785-1KG BT224785-250G
Strontium carbonate ≥99%
Strontium carbonate

CAS: 1633-05-2; EC No: 216-643-7; MDL No: MFCD00011250 -325 Mesh Powder, typically 1 micron; Linear Formula: SrCO3; MW: 147.63 Melting Point: 1100° (decomposes) Density (g/mL): 3.7


  • -325 Mesh Powder, typically 1 micron

Formula: CO₃Sr
Melting Pt: 1100 °C (dec.)
Density: 3.7 g/mL
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR