4-Hydroxyphenylboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: (4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid

224610-1G 224610-5G
BT224610-1GEA 51 USD
BT224610-1G BT224610-5G
4-Hydroxyphenylboronic acid ≥97%
4-Hydroxyphenylboronic acid

CAS: 71597-85-8; MDL No: MFCD01074628 Solid; Molecular Formula: C6H7BO3; MW: 137.93


  • Solid

Formula: C₆H₇BO₃
MW: 137.93 g/mol
MDL Number: MFCD01074628
CAS Number: 71597-85-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR