Polyimide, resin
Supplier: BeanTown Chemical
223720-10G
223720-50G
BT223720-10GEA
63.95
USD
BT223720-10G
BT223720-50G
Polyimide, resin
Polyimide
CAS: 62929-02-6; MDL No: MFCD01868102
Powder
Melting Point:
- Powder
Formula:
C₃₅H₂₈N₂O₇ Melting Pt: >300 °C Density: 1.2 g/mL Flash Pt: 86 °C (197 °F) |
MDL Number:
MFCD01868102 CAS Number: 62929-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...