1-Butanesulfonic acid sodium salt ≥99%
Supplier: BeanTown Chemical
Synonyms:
Sodium 1-butanesulphonate, Sodium butane-1-sulphonate, Sodium 1-butanesulfonate, Butane-1-sulfonic acid sodium salt, Sodium 1-butansulphonate, Sodium 1-butansulfonate, Butane-1-sulphonic acid sodium salt, Sodium butane-1-sulfonate
223545-100G
223545-25G
223545-5G
BT223545-100GEA
557.05
USD
BT223545-100G
BT223545-25G
BT223545-5G
1-Butanesulfonic acid sodium salt ≥99%
1-Butansulphonic acid sodium salt
CAS: 2386-54-1; EC No: 219-201-1; MDL No: MFCD00007540
Crystalline; Linear Formula: CH3(CH2)3SO3Na; Molecular Formula: C4H9NaO3S; MW: 160.17
Melting Point:
- Crystalline
Formula:
C₄H₉NaO₃S MW: 160.17 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00007540 CAS Number: 2386-54-1 EINECS: 219-201-1 |
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...