Lutetium ≥99.9% (trace metals basis excluding Ta), rods Ø 6.35 mm
Supplier: BeanTown Chemical
223360-10MM
223360-50MM
BT223360-10MMEA
493
USD
BT223360-10MM
BT223360-50MM
Lutetium ≥99.9% (trace metals basis excluding Ta), rods Ø 6.35 mm
Lutetium
CAS: 7439-94-3; EC No: 231-103-0; MDL No: MFCD00011098
Solid, cast rod, diameter is approximate; Molecular Formula: Lu; MW: 174.97
Melting Point: 1663°; Boiling Point: 3402°
Density (g/mL): 9.84
- Solid, cast rod, diameter is approximate
Formula:
Lu Boiling Pt: 3402 °C Melting Pt: 1663 °C Density: 9.84 g/mL |
MDL Number:
MFCD00011098 CAS Number: 7439-94-3 EINECS: 231-103-0 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...