1H-Pyrazole-1-carboximidamide hydrochloride ≥95%

Supplier: BeanTown Chemical

Synonyms: 1-Amidinopyrazole Hydrochloride, 1H-Pyrazole-1-carboxamidine monohydrochloride, Hydrochloride, 1-Pyrazolecarboxamidine, Praxadine

223350-10G 223350-50G
BT223350-10GEA 47 USD
BT223350-10G BT223350-50G
1H-Pyrazole-1-carboximidamide hydrochloride ≥95%
1H-Pyrazole-1-carboximidamide hydrochloride

CAS: 4023-02-3; EC No: 429-520-1; MDL No: MFCD00210087 Crystalline; Molecular Formula: C4H6N4·HCl; MW: 146.58 Melting Point: 167-170°


  • Crystalline

Formula: C₄H₇ClN₄
MW: 146.58 g/mol
Melting Pt: 167…170 °C
MDL Number: MFCD00210087
CAS Number: 4023-02-3
EINECS: 429-520-1

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR