Citric acid ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

222995-1KG 222995-250G 222995-5KG
BT222995-1KGEA 56.65 USD
BT222995-1KG BT222995-250G BT222995-5KG
Citric acid ≥99%
Citric acid

CAS: 77-92-9; EC No: 201-069-1; MDL No: MFCD00011669; RTECS: GE7350000 Crystalline; Molecular Formula: C6H8O7; MW: 192.13 Melting Point: 153-159° Hygroscopic


  • RTECS: GE7350000
  • Hygroscopic
  • Crystalline

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Melting Pt: 153…159 °C
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR