Dysprosium(III) oxide ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Didysprosium trioxide, Dysprosium oxide

222585-100G 222585-25G 222585-500G
BT222585-100GEA 479 USD
BT222585-100G BT222585-25G BT222585-500G
Dysprosium(III) oxide ≥99.99%
Dysprosium (III) oxide

CAS: 1308-87-8; EC No: 215-164-0; MDL No: MFCD00010986 Powder; Molecular Formula: Dy2O3; MW: 373.00 Density (g/mL): 7.81 Hygroscopic


  • Hygroscopic
  • Powder

Formula: Dy₂O₃
MW: 373 g/mol
Melting Pt: 2400
Density: 7.81 g/mL
MDL Number: MFCD00010986
CAS Number: 1308-87-8
EINECS: 215-164-0
Merck Index: 14,03482

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR