Tungstosilicic acid hydrate

Supplier: BeanTown Chemical
Warning

Synonyms: Silicotungstic acid

222405-10G 222405-50G
BT222405-10GEA 31.86 USD
BT222405-10G BT222405-50G
Tungstosilicic acid hydrate
Tungstosilicic acid Hydrate

CAS: 12027-43-9; MDL No: MFCD00036271 Crystalline; Linear Formula: H4SiO4·12WO3·xH2O; MW: 2878.28 (anhydrous)


  • Crystalline

Formula: H₄Si(W₃O₁₀)₄.xH₂O
MW: 2896.172 g/mol
MDL Number: MFCD00036271
CAS Number: 12027-43-9
Merck Index: 14,8502

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR