3-(3,4-Dihydroxyphenyl)-L-alanine ≥98%

Supplier: BeanTown Chemical

Synonyms: Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa

222035-100G 222035-25G 222035-5G
BT222035-100GEA 384 USD
BT222035-100G BT222035-25G BT222035-5G
3-(3,4-Dihydroxyphenyl)-L-alanine ≥98%
3-(3,4-Dihydroxyphenyl)-L-alanine

CAS: 59-92-7; EC No: 200-445-2; MDL No: MFCD00002598; RTECS: AY5600000 Crystalline; Molecular Formula: C9H11NO4; MW: 197.19 Melting Point: 276-278°


  • RTECS: AY5600000
  • Crystalline

Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
Melting Pt: 276…278 °C
MDL Number: MFCD00002598
CAS Number: 59-92-7
EINECS: 200-445-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR