Strontium carbonate ≥99.995%

Supplier: BeanTown Chemical

221630-100G 221630-25G 221630-500G
BT221630-100GEA 388 USD
BT221630-100G BT221630-25G BT221630-500G
Strontium carbonate ≥99.995%
Strontium carbonate

CAS: 1633-05-2; EC No: 216-643-7; MDL No: MFCD00011250 -22 Mesh Powder; Linear Formula: SrCO3; MW: 147.63 Melting Point: 1100° (decomposes) Density (g/mL): 3.70


  • -22 Mesh Powder

Formula: CO₃Sr
Melting Pt: 1100 °C (dec.)
Density: 3.70 g/mL
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR