2-Methoxy-3-nitropyridine ≥98%

Supplier: BeanTown Chemical

221550-1G 221550-25G 221550-5G
BT221550-1GEA 37 USD
BT221550-1G BT221550-25G BT221550-5G
2-Methoxy-3-nitropyridine ≥98%
2-Methoxy-3-nitropyridine

CAS: 20265-35-4; MDL No: MFCD00023459 Solid; Molecular Formula: C6H6N2O3; MW: 154.12 Melting Point: 52-57°


  • Solid

Formula: C₆H₆N₂O₃
Melting Pt: 52…57 °C
MDL Number: MFCD00023459
CAS Number: 20265-35-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR