Sorbic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
(E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
221410-1KG
221410-250G
BT221410-1KGEA
89.01
USD
BT221410-1KG
BT221410-250G
Sorbic acid ≥99%
Sorbic acid
CAS: 110-44-1; EC No: 203-768-7; MDL No: MFCD00002703; RTECS: WG2100000
Crystalline; Linear Formula: CH2CH=CHCH=CHCOOH; Molecular Formula: C6H8O2; MW: 112.13
Melting Point: 132-135°; Boiling Point: 170° (decomposes); Flash point: 127°C (261°F)
Density (g/mL): 1.2
- RTECS: WG2100000
- Crystalline
Formula:
CH₃CH=CHCH=CHCOOH MW: 112.13 g/mol Boiling Pt: 228 (dec.) Melting Pt: 132…135 °C Density: 1.2 g/mL Flash Pt: 127 °C (261 °F) |
MDL Number:
MFCD00002703 CAS Number: 110-44-1 EINECS: 203-768-7 Merck Index: 13,08793 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...