Biochanin A ≥98%

Supplier: BeanTown Chemical

Synonyms: 5,7-Dihydroxy-3-(4-methoxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-methoxyphenyl)chromen-4-one, 4'-Methoxy-5,7-dihydroxyisoflavone, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-4'-methoxyisoflavone

220480-1G 220480-250MG 220480-5G
BT220480-1GEA 177 USD
BT220480-1G BT220480-250MG BT220480-5G
Biochanin A ≥98%
Biochanin A

CAS: 491-80-5; EC No: 207-744-7; MDL No: MFCD00006839 Powder; Molecular Formula: C16H12O5; MW: 284.26 Melting Point: 210-213°


  • Powder

Formula: C₁₆H₁₂O₅
Melting Pt: 210…213 °C
MDL Number: MFCD00006839
CAS Number: 491-80-5
EINECS: 207-744-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR