Calmagite

Supplier: BeanTown Chemical

220455-25G 220455-5G
BT220455-25GEA 93.62 USD
BT220455-25G BT220455-5G
Calmagite
Calmagite

CAS: 3147-14-6; EC No: 221-563-0; MDL No: MFCD00011656 Powder; Molecular Formula: C17H14N2O5S; MW: 358.37 Melting Point: 330°


  • Powder

Formula: C₁₇H₁₄N₂O₅S
MW: 358.37
Melting Pt: 330 °C
MDL Number: MFCD00011656
CAS Number: 3147-14-6
EINECS: 221-563-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR