Silver ≥99.9% (trace metals basis), wire Ø 0.5 mm, half hard type
Supplier: BeanTown Chemical
220275-10M
220275-2M
220275-50M
BT220275-10MEA
145.17
USD
BT220275-10M
BT220275-2M
BT220275-50M
Silver ≥99.9% (trace metals basis), wire Ø 0.5 mm, half hard type
Silver
CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397; RTECS: VW3500000
Solid; Molecular Formula: Ag; MW: 107.87
Melting Point: 960°; Boiling Point: 2212°
Density (g/mL): 10.49
- RTECS: VW3500000
- Solid
Formula:
Ag Boiling Pt: 2212 °C Melting Pt: 960 °C Density: 10.49 g/mL |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...