2',3'-O-Isopropylideneadenosine ≥98%

Supplier: BeanTown Chemical

219480-25G 219480-5G
BT219480-25GEA 187 USD
BT219480-25G BT219480-5G
2',3'-O-Isopropylideneadenosine ≥98%
2',3'-O-Isopropylideneadenosine

CAS: 362-75-4; EC No: 206-650-3; MDL No: MFCD00005756 Crystalline; Molecular Formula: C13H17N5O4; MW: 307.31 Melting Point: 221-222° Optical Rotation: [α]20/D −98.5°, c = 1 in dioxane


  • Optical rotation: [α]20/D −98.5°, c = 1 in dioxane
  • Crystalline

Formula: C₁₃H₁₇N₅O₄
Melting Pt: 221…222 °C
MDL Number: MFCD00005756
CAS Number: 362-75-4
EINECS: 206-650-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR