2,1,3-Benzothiadiazole ≥98%

Supplier: BeanTown Chemical

219140-25G 219140-5G
BT219140-25GEA 123 USD
BT219140-25G BT219140-5G
2,1,3-Benzothiadiazole ≥98%
2,1,3-Benzothiadiazole

CAS: 273-13-2; EC No: 205-985-2; MDL No: MFCD00005809 Powder; Molecular Formula: C6H4N2S; MW: 136.17 Melting Point: 42-44°; Boiling Point: 206°; Flash point: 95°C (203°F)


  • Powder

Formula: C₆H₄N₂S
Boiling Pt: 206 °C
Melting Pt: 42…44 °C
Flash Pt: 95 °C (203 °F)
MDL Number: MFCD00005809
CAS Number: 273-13-2
EINECS: 205-985-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR